Abnova
Manufacturer Code:PAB6174
Catalog # 89-132-211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to human GULP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of GULP1.
For Research Use Only
Protein Aliases: CED-6; CED6; Cell death protein 6 homolog; engulfment adapter protein; FLJ31156; GULP; Protein GULP; PTB domain adapter protein CED-6; PTB domain adaptor protein CED-6; PTB domain-containing engulfment adapter protein 1
Gene Aliases: CED-6; CED6; GULP; GULP1
UniProt ID: (Human) Q9UBP9
Entrez Gene ID: (Human) 51454
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.