Abnova
Manufacturer Code:H00051454A01
Catalog # 89-106-905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. to 100 a.a) partial recombinant protein with GST tag.,GULP1 (AAH01103 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant GULP1.
For Research Use Only
Protein Aliases: CED-6; CED6; Cell death protein 6 homolog; engulfment adapter protein; FLJ31156; GULP; Protein GULP; PTB domain adapter protein CED-6; PTB domain adaptor protein CED-6; PTB domain-containing engulfment adapter protein 1
Gene Aliases: CED-6; CED6; GULP; GULP1
UniProt ID: (Human) Q9UBP9
Entrez Gene ID: (Human) 51454
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.