Invitrogen
Manufacturer Code:PA570006
Catalog # PIPA570006
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human GUK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP:GMP phosphotransferase; ATP:GMP-phosphotransferase; GMP kinase; Guanylate kinase; Guanylate kinase 1; RP11-520H14.2 GMK ATP:GMP phosphotransferase GMP kinase guanylate kinase
Gene Aliases: AA409738; AL033299; AL033300; AV026611; GMK; GMPK; GUK; Guk-1; GUK1
UniProt ID: (Human) Q16774, (Bovine) P46195, (Mouse) Q64520
Entrez Gene ID: (Dog) 475178, (Rabbit) 100354087, (Human) 2987, (Bovine) 281217, (Horse) 100147433, (Mouse) 14923, (Guinea pig) 100733447, (Rat) 303179
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.