Proteintech
Manufacturer Code:107551AP150UL
Catalog # 50-172-6569
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GUK1 Fusion Protein Ag1205 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP:GMP phosphotransferase; ATP:GMP-phosphotransferase; GMK GMP kinase Guanylate kinase guanylate kinase 1 GUK1; GMP kinase; Guanylate kinase; Guanylate kinase 1
Gene Aliases: AA409738; AL033299; AL033300; AV026611; GMK; GMPK; Guk-1; GUK1
UniProt ID: (Human) Q16774, (Mouse) Q64520
Entrez Gene ID: (Human) 2987, (Mouse) 14923, (Rat) 303179
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.