Proteintech
Manufacturer Code:174891AP150UL
Catalog # 50-173-1600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GUF1 Fusion Protein Ag11579 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EF-4; Elongation factor 4 homolog; GTP-binding protein GUF1 homolog; GTPase GUF1; GTPase of unknown function 1; GUF1; GUF1 GTPase homolog; Ribosomal back-translocase; Translation factor GUF1, mitochondrial
Gene Aliases: 4631409J12; AA407526; EF-4; GUF1
UniProt ID: (Human) Q8N442, (Mouse) Q8C3X4
Entrez Gene ID: (Human) 60558, (Mouse) 231279
Molecular Function:
G-protein
RNA binding protein
enzyme modulator
hydrolase
nucleic acid binding
translation elongation factor
translation factor
translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.