Invitrogen
Manufacturer Code:PA542803
Catalog # PIPA542803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GUCY1B3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta 1 sGC; GCS-beta-1; GCS-beta-3; Guanylate cyclase 1 soluble beta 3 GC-S-beta-1 GC-SB3 GUC1B3 GUCB3 GUCSB3 GUCY1B1; guanylate cyclase 1, soluble, beta 3; Guanylate cyclase soluble subunit beta-1; Guanylate cyclase soluble subunit beta-3; guanylate cyclase, soluble, beta 1; nitric oxide-sensitive guanylyl cyclase beta 1 subunit; NO-sensitive GC beta 1 subunit; NO-sensitive guanylyl cyclase beta 1 subunit; soluble guanylate cyclase 1 beta 3; soluble guanylate cyclase beta-1 subunit; Soluble guanylate cyclase small subunit; soluble guanylate cyclase small subunit beta 1; soluble guanylyl cyclase beta1 subunit
Gene Aliases: GC-S-beta-1; GC-SB3; GCbeta1; GUC1B3; GUCB3; GUCSB3; GUCY1B1; GUCY1B3; SGC; sgc1b1; si:ch73-34b5.3
UniProt ID: (Bovine) P16068, (Dog) Q4ZHR9, (Human) Q02153, (Rat) P20595, (Mouse) O54865
Entrez Gene ID: (Rabbit) 100349851, (Horse) 100069551, (Bovine) 282433, (Dog) 553102, (Human) 2983, (Guinea pig) 100723676, (Rat) 25202, (Mouse) 54195, (Zebrafish) 100150304
Molecular Function:
adenylate cyclase
cyclase
guanylate cyclase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.