Abnova
Manufacturer Code:PAB4059
Catalog # 89-131-256
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human GUCY1B3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GUCY1B3.
For Research Use Only
Protein Aliases: GC-S-beta-1; GC-SB3; GCS-beta-1; GCS-beta-3; guanylate cyclase 1, soluble, beta 3; Guanylate cyclase soluble subunit beta-1; Guanylate cyclase soluble subunit beta-3; GUC1B3; GUCB3; GUCSB3; GUCY1B1; Soluble guanylate cyclase small subunit
Gene Aliases: GC-S-beta-1; GC-SB3; GUC1B3; GUCB3; GUCSB3; GUCY1B1; GUCY1B3
UniProt ID: (Human) Q02153
Entrez Gene ID: (Human) 2983
Molecular Function:
adenylate cyclase
cyclase
guanylate cyclase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.