Proteintech
Manufacturer Code:190111AP150UL
Catalog # 50-173-2320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GUCY1B3 Fusion Protein Ag5196 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta 1 sGC; GC S beta 1 GC SB3 GCS beta 1 GCS beta 3 GUC1B3 GUCB3 GUCSB3 GUCY1B1 GUCY1B3; GCS-beta-1; GCS-beta-3; guanylate cyclase 1, soluble, beta 3; Guanylate cyclase soluble subunit beta-1; Guanylate cyclase soluble subunit beta-3; guanylate cyclase, soluble, beta 1; NO-sensitive guanylyl cyclase beta 1 subunit; soluble guanylate cyclase beta-1 subunit; Soluble guanylate cyclase small subunit
Gene Aliases: GC-S-beta-1; GC-SB3; GCbeta1; GUC1B3; GUCB3; GUCSB3; GUCY1B1; GUCY1B3; SGC
UniProt ID: (Human) Q02153, (Rat) P20595, (Mouse) O54865
Entrez Gene ID: (Human) 2983, (Rat) 25202, (Mouse) 54195
Molecular Function:
adenylate cyclase
cyclase
guanylate cyclase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.