Invitrogen
Manufacturer Code:MA517086
Catalog # PIMA517086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3G6B2 |
Immunogen | Purified recombit fragment of human GUCY1A3 (amino acids: 22-214) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GC-S-alpha-1; GCS-alpha-1; GCS-alpha-3; guanylate cyclase 1, soluble, alpha 3; Guanylate cyclase soluble subunit alpha-1; Guanylate cyclase soluble subunit alpha-3; Soluble guanylate cyclase large subunit
Gene Aliases: GC-SA3; GUC1A3; GUCA3; GUCSA3; GUCY1A1; GUCY1A3; MYMY6
UniProt ID: (Human) Q02108
Entrez Gene ID: (Human) 2982
Molecular Function:
adenylate cyclase
cyclase
guanylate cyclase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.