Novus Biologicals
Manufacturer Code:NBP15806620UL
Catalog # NBP15806620
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to GTSE1(G-2 and S-phase expressed 1) The peptide sequence was selected from the N terminal of GTSE1. Peptide sequence NNPVPEQPPLPTSESPFAWSPLAGEKFVEVYKEAHLLALHIESSSRNQAA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B99 G2 and S phase-expressed protein 1 G-2 and S-phase expressed 1 GTSE-1 Protein B99 homolog; G-2 and S-phase expressed 1; G2 and S phase-expressed protein 1; GTSE-1; Protein B99 homolog
Gene Aliases: B99; GTSE1
UniProt ID: (Human) Q9NYZ3
Entrez Gene ID: (Human) 51512
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.