Invitrogen
Manufacturer Code:PA541840
Catalog # PIPA541840
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GTPBP9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA damage-regulated overexpressed in cancer 45; DOC45; GTP-binding protein 9; GTP-binding protein 9 (putative); GTP-binding protein PTD004; Obg-like ATPase 1; obg-like ATPase 1 {ECO:0000255|HAMAP-Rule:MF_03167}; obg-like ATPase 1-like protein
Gene Aliases: 2510025G09Rik; 2810405J23Rik; 2810409H07Rik; DOC45; GBP45; GTBP9; GTPBP9; OLA1; PRO2455; PTD004; RGD1307982
UniProt ID: (Human) Q9NTK5, (Bovine) Q2HJ33, (Rat) A0JPJ7, (Mouse) Q9CZ30
Entrez Gene ID: (Human) 29789, (Bovine) 509966, (Dog) 478803, (Rabbit) 100349489, (Horse) 100053204, (Rat) 296488, (Mouse) 67059, (Guinea pig) 100734863
Molecular Function:
G-protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.