Novus Biologicals
Manufacturer Code:NBP15745320UL
Catalog # NBP15745320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to GTPBP9 (GTP-binding protein 9 (putative)) The peptide sequence was selected from the N terminal of GTPBP9. Peptide sequence MIGPIIDKLEKVAVRGGDKKLKPEYDIMCKVKSWVIDQKKPVRFYHDWND. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp313H1942 DNA damage-regulated overexpressed in cancer 45 protein DOC45 EC 3.6.3 EC 3.6.3.- GBP45 GTP-binding protein 9 GTP-binding protein 9 (putative) GTP-binding protein PTD004 GTPBP9 homologous yeast-44.2 protein Obg-like ATPase 1 PTD004; DNA damage-regulated overexpressed in cancer 45; DOC45; GTP-binding protein 9; Obg-like ATPase 1
Gene Aliases: DOC45; GBP45; GTBP9; GTPBP9; OLA1; PRO2455; PTD004
UniProt ID: (Human) Q9NTK5
Entrez Gene ID: (Human) 29789
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.