Proteintech
Manufacturer Code:201331AP150UL
Catalog # 50-173-2720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GTPBP5 Fusion Protein Ag14015 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP binding protein 5; GTP binding protein 5 (putative); GTP binding protein 5 GTPBP5 ObgH1 Protein obg homolog 1; GTP-binding protein 5; GTP-binding protein 5 (putative); Mitochondrial ribosome-associated GTPase 2; ObgH1; Protein obg homolog 1
Gene Aliases: 1810011P19Rik; 2900056P18Rik; D2Bwg0647e; dJ1005F21.2; GTPBP5; MTG2; OBGH1
UniProt ID: (Human) Q9H4K7
Entrez Gene ID: (Human) 26164, (Mouse) 52856, (Rat) 296462
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.