Novus Biologicals
Manufacturer Code:NBP216765
Catalog # NBP216765
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human GTPBP3. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ14700 FLJ17836 GTP binding protein 3 (mitochondrial) GTPBG3 GTP-binding protein 3 MSS1 MTGP1Mitochondrial GTP-binding protein 1 THDF1 tRNA modification GTPase GTPBP3 mitochondrial; GTP-binding protein 3; Mitochondrial GTP-binding protein 1; tRNA modification GTPase GTPBP3, mitochondrial
Gene Aliases: COXPD23; GTPBG3; GTPBP3; MSS1; MTGP1; THDF1
UniProt ID: (Human) Q969Y2
Entrez Gene ID: (Human) 84705
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.