Proteintech
Manufacturer Code:107641AP150UL
Catalog # 50-172-6577
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GTPBP3 Fusion Protein Ag1198 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP binding protein 3 (mitochondrial); GTP binding protein 3 GTPBG3 GTPBP3 MSS1 MTGP1 THDF1; GTP-binding protein 3; Mitochondrial GTP-binding protein 1; tRNA modification GTPase GTPBP3, mitochondrial
Gene Aliases: 2410009F13Rik; AI607903; COXPD23; GTPBG3; GTPBP3; MSS1; MTGP1; THDF1
UniProt ID: (Human) Q969Y2, (Mouse) Q923K4, (Rat) Q5PQQ1
Entrez Gene ID: (Human) 84705, (Mouse) 70359, (Rat) 290633
Molecular Function:
G-protein
enzyme modulator
small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.