Novus Biologicals
Manufacturer Code:NBP15764820UL
Catalog # NBP15764820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to GTPBP2(GTP binding protein 2) The peptide sequence was selected from the N terminal of GTPBP2. Peptide sequence GCGGPKGKKKNGRNRGGKANNPPYLPPEAEDGNIEYKLKLVNPSQYRFEH. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GTP binding protein 2 GTP-binding protein 2 MGC74725; GTP-binding protein 2; GTPBP2
Gene Aliases: GTPBP2
UniProt ID: (Human) Q9BX10
Entrez Gene ID: (Human) 54676
Molecular Function: G-protein RNA binding protein enzyme modulator hydrolase nucleic acid binding translation elongation factor translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.