Proteintech
Manufacturer Code:163741AP150UL
Catalog # 50-173-0902
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GTPBP1 Fusion Protein Ag9332 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein 1 GP 1 GP1 GTP binding protein 1 GTPBP1 HSPC018; G-protein 1; GP1; GTP-binding protein 1
Gene Aliases: AL022987; AW045683; C85212; GP-1; GP1; Gtpbp; GTPBP1; HSPC018
UniProt ID: (Human) O00178, (Rat) D2XV59, (Mouse) O08582
Entrez Gene ID: (Human) 9567, (Rat) 300077, (Mouse) 14904
Molecular Function: G-protein RNA binding protein enzyme modulator hydrolase nucleic acid binding translation elongation factor translation factor translation initiation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.