Invitrogen
Manufacturer Code:PA577132
Catalog # PIPA577132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human C16orf80. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basal body up-regulated protein 22; Cilia- and flagella-associated protein 20; Cilia- and flagella-associated protein 20 Basal body up-regulated protein 22 Transcription factor IIB CFAP20 BUG22 C16orf80; flagellar associated protein 20 homolog; functional spliceosome-associated protein 23; gene trap locus 3; Gene trap locus 3 protein; Gene trap locus 3 protein homolog; Transcription factor IIB; UPF0468 protein C16orf80; UPF0468 protein C16orf80 homolog
Gene Aliases: 2600014O15Rik; AL024127; BUG22; C16orf80; CFAP20; EVORF; fSAP23; GTL3; T10-2A2
UniProt ID: (Human) Q9Y6A4, (Mouse) Q8BTU1, (Rat) Q499T7
Entrez Gene ID: (Human) 29105, (Mouse) 14894, (Rat) 307642
Molecular Function:
nuclease
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.