OriGene
Manufacturer Code:CF812628
Catalog # 50-167-0962
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4A1 |
Immunogen | Human recombit protein fragment corresponding to amino acids 583-822 of human GTF3C4 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: general transcription factor 3C 4; General transcription factor 3C polypeptide 4; general transcription factor IIIC, polypeptide 4, 90kDa; KAT12 TF3C-delta TFIII90 TFIIIC290 TFIIIC90 TFIIICDELTA; TF3C-delta; TFIIIC 90 kDa subunit; Transcription factor IIIC 90 kDa subunit; Transcription factor IIIC subunit delta
Gene Aliases: GTF3C4; KAT12; TF3C-delta; TFIII90; TFIIIC290; TFIIIC90; TFIIICDELTA
UniProt ID: (Human) Q9UKN8
Entrez Gene ID: (Human) 9329
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.