Proteintech
Manufacturer Code:176531AP150UL
Catalog # 50-173-1704
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GTF3C4 Fusion Protein Ag11898 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: general transcription factor 3C 4; General transcription factor 3C polypeptide 4; general transcription factor IIIC 4; general transcription factor IIIC, polypeptide 4, 90kDa; GTF3C4 KAT12 TF3C delta TFIII90 TFIIIC 90 kDa subunit TFiiiC2 90 TFIIIC90 TFIIICdelta; TF3C-delta; TFIIIC 90 kDa subunit; Transcription factor IIIC 90 kDa subunit; Transcription factor IIIC subunit delta
Gene Aliases: 5330400C03; AI426938; AU014771; AU017413; GTF3C4; KAT12; TF3C-delta; TFIII90; TFIIIC290; TFIIIC90; TFIIICDELTA
UniProt ID: (Human) Q9UKN8, (Mouse) Q8BMQ2
Entrez Gene ID: (Human) 9329, (Mouse) 269252, (Rat) 685539
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.