Invitrogen
Manufacturer Code:PA541578
Catalog # PIPA541578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GTF2IRD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: binding factor for early enhancer; general transcription factor 3; general transcription factor II I repeat domain-containing 1; General transcription factor II-I repeat domain-containing protein 1; General transcription factor III; GTF2I repeat domain containing 1 CREAM1 GTF3 MUSTRD1 RBAP2 WBSCR11 WBSCR12 hMusTRD1alpha1 BEN WBS; GTF2I repeat domain-containing protein 1; Muscle TFII-I repeat domain-containing protein 1; muscle TFII-I repeat domain-containing protein 1 alpha 1; MusTRD1/BEN; Slow-muscle-fiber enhancer-binding protein; USE B1-binding protein; Williams-Beuren syndrome chromosomal region 11 protein; Williams-Beuren syndrome chromosomal region 12 protein; Williams-Beuren syndrome chromosome region 11
Gene Aliases: BEN; CREAM1; GTF2IRD1; GTF3; hMusTRD1alpha1; MUSTRD1; RBAP2; WBS; WBSCR11; WBSCR12
UniProt ID: (Human) Q9UHL9
Entrez Gene ID: (Bovine) 507792, (Rabbit) 100340448, (Pig) 100510956, (Human) 9569, (Horse) 100062277, (Dog) 489796, (Rat) 246770, (Guinea pig) 100726332
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.