Proteintech
Manufacturer Code:170521AP150UL
Catalog # 50-173-1351
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GTF2IRD1 Fusion Protein Ag10718 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alb-c-myc line 166.8; Alb/c-myc line 166.8; BEN CREAM1 GTF2IRD1 GTF3 hMusTRD1alpha1 MUSTRD1 MusTRD1/BEN RBAP2 USE B1 binding protein WBS WBSCR11 WBSCR12; Binding factor for early enhancer; c-myc line 166.8; general transcription factor 3; general transcription factor II I repeat domain-containing 1; General transcription factor II-I repeat domain-containing protein 1; General transcription factor III; GTF2I repeat domain-containing 1; GTF2I repeat domain-containing protein 1; Muscle TFII-I repeat domain-containing protein 1; muscle TFII-I repeat domain-containing protein 1 alpha 1; MusTRD1/BEN; Slow-muscle-fiber enhancer-binding protein; transcription factor GTF3 alpha 2; transcription factor GTF3 gamma 2; USE B1-binding protein; Williams-Beuren syndrome chromosomal region 11 protein; Williams-Beuren syndrome chromosomal region 12 protein; Williams-Beuren syndrome chromosome region 11
Gene Aliases: 1700012P16Rik; BEN; CREAM1; ESTM9; Gtf2il; GTF2IRD1; GTF3; hMusTRD1alpha1; MUSTRD1; RBAP2; Tg(Alb1-Myc)166.8Sst; WBS; WBSCR11; WBSCR12; X83320
UniProt ID: (Human) Q9UHL9, (Mouse) Q9JI57
Entrez Gene ID: (Human) 9569, (Mouse) 57080, (Rat) 246770
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.