Invitrogen
Manufacturer Code:PA5117110
Catalog # PIPA5117110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human GTF2I |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6030441I21Rik BAP135 BAP-135 Bruton tyrosine kinase-associated protein 135 BTKAP1 BTK-associated protein 135 BTK-associated protein 135kD DIWS Diws1t general transcription factor II I general transcription factor IIi General transcription factor II-I Gtf2i Gtf2ird1 GTFII-I IB291 OTTHUMP00000209509 SPIN SRF-Phox1-interacting protein tfiii TFII-I WBS WBSCR6 Williams-Beuren syndrome chromosomal region 6 protein Williams-Beuren syndrome chromosome region 6; BAP-135; Bruton tyrosine kinase-associated protein 135; BTK-associated protein 135; BTK-associated protein, 135kD; General transcription factor II-I; GTFII-I; SPIN; SRF-Phox1-interacting protein; TFII-I; Williams-Beuren syndrome chromosomal region 6 protein; Williams-Beuren syndrome chromosome region 6
Gene Aliases: 6030441I21Rik; BAP-135; BAP135; BTKAP1; DIWS; Diws1t; GTF2I; Gtf2ird1; GTFII-I; IB291; SPIN; TFII-I; WBS; WBSCR6
UniProt ID: (Human) P78347, (Rat) Q5U2Y1, (Mouse) Q9ESZ8
Entrez Gene ID: (Human) 2969, (Rat) 353256, (Mouse) 14886
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.