Invitrogen
Manufacturer Code:PA540374
Catalog # PIPA540374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GTF2H3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic transcription factor 2 34 kDa subunit; BTF2 p34; BTF2-p34; General transcription factor IIH BTF2 TFB4 TFIIH; General transcription factor IIH polypeptide 3; General transcription factor IIH subunit 3; general transcription factor IIH, polypeptide 3, 34kDa; TFIIH basal transcription factor complex p34 subunit
Gene Aliases: 34kDa; 5033417D07Rik; BTF2; BTF2 p34; C730029A10; D5Ertd679e; GTF2H3; P34; TFB4; TFIIH
UniProt ID: (Human) Q13889, (Bovine) Q05B56, (Mouse) Q8VD76, (Rat) Q561R7
Entrez Gene ID: (Rabbit) 100347110, (Human) 2967, (Pig) 100152121, (Bovine) 511896, (Horse) 100059174, (Mouse) 209357, (Rat) 288651, (Guinea pig) 100720023
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.