Proteintech
Manufacturer Code:151391AP150UL
Catalog # 50-173-0014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GTF2F2 Fusion Protein Ag7313 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP dependent helicase GTF2F2 BTF4 GTF2F2 RAP30 TF2F2 TFIIF TFIIF beta; ATP-dependent helicase GTF2F2; General transcription factor IIF 30 kDa subunit; General transcription factor IIF subunit 2; general transcription factor IIF, polypeptide 2 (30kD subunit); general transcription factor IIF, polypeptide 2, 30kDa; TFIIF-beta; transcription factor gamma; Transcription initiation factor IIF subunit beta; Transcription initiation factor RAP30
Gene Aliases: 1110031C13Rik; AI326315; BTF4; GTF2F2; RAP30; TF2F2; TFIIF
UniProt ID: (Human) P13984, (Mouse) Q8R0A0
Entrez Gene ID: (Human) 2963, (Mouse) 68705, (Rat) 81674
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.