Invitrogen
Manufacturer Code:MA525660
Catalog # PIMA525660
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D5 |
Immunogen | Full length human recombit protein of GTF2F1 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BTF4 C76800 General transcription factor IIF 74 kDa subunit General transcription factor IIF subunit 1 General transcription factor IIF polypeptide 1 74kDa Gtf2f1 MGC94148 OTTHUMP00000237859 RAP74 T2FA&lowbarHUMAN TF2F1 TFIIF TFIIF-alpha Transcription initiation factor IIF subunit alpha Transcription initiation factor RAP74; General transcription factor IIF 74 kDa subunit; General transcription factor IIF subunit 1; general transcription factor IIF, polypeptide 1, 74kDa; TFIIF-alpha; Transcription initiation factor IIF subunit alpha; Transcription initiation factor RAP74
Gene Aliases: BTF4; GTF2F1; RAP74; TF2F1; TFIIF
UniProt ID: (Human) P35269
Entrez Gene ID: (Human) 2962
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.