Invitrogen
Manufacturer Code:PA5109796
Catalog # PIPA5109796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human GTF2F1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810405L04Rik BTF4 C76800 General transcription factor IIF 74 kDa subunit General transcription factor IIF subunit 1 GTF2F1 MGC94148 OTTHUMP00000237859 RAP74 TF2F1 TFIIF TFIIF-alpha Transcription initiation factor IIF subunit alpha Transcription initiation factor RAP74; General transcription factor IIF 74 kDa subunit; general transcription factor IIF polypeptide 1 (74kD subunit); General transcription factor IIF subunit 1; general transcription factor IIF, polypeptide 1, 74kDa; TFIIF-alpha; Transcription initiation factor IIF subunit alpha; Transcription initiation factor RAP74
Gene Aliases: 2810405L04Rik; BTF4; C76800; GTF2F1; RAP74; TF2F1; TFIIF
UniProt ID: (Human) P35269, (Rat) Q6AY96, (Mouse) Q3THK3
Entrez Gene ID: (Human) 2962, (Rat) 316123, (Mouse) 98053
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.