Invitrogen
Manufacturer Code:PA540504
Catalog # PIPA540504
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GSH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Genetic-screened homeobox 2; genomic screened homeo box 2; genomic screened homeo box 2 protein; GS homeobox 2; GS homeobox 2 GSH2; Homeobox protein GSH-2
Gene Aliases: Gsh-2; GSH2; GSX2
UniProt ID: (Human) Q9BZM3, (Mouse) P31316
Entrez Gene ID: (Human) 170825, (Dog) 482152, (Bovine) 539228, (Rabbit) 100340797, (Horse) 100054004, (Pig) 100524448, (Mouse) 14843, (Rat) 364140
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.