Invitrogen
Manufacturer Code:PA552088
Catalog # PIPA552088
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GSTZ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; Glutathione S-transferase zeta 1; glutathione transferase zeta 1; GSTZ1-1; GSTZ1-1 MAAI MAI S-(hydroxyalkyl)glutathione lyase glutathione S-alkyltransferase glutathione S-aralkyltransferase glutathione S-aryltransferase glutathione s-transferase Zeta 1 maleylacetoacetate isomerase maleylacetone isomerase; MAAI; Maleylacetoacetate isomerase; maleylacetone isomerase; S-(hydroxyalkyl)glutathione lyase
Gene Aliases: GSTZ1; GSTZ1-1; MAAI; MAI
UniProt ID: (Human) O43708
Entrez Gene ID: (Human) 2954
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.