Invitrogen
Manufacturer Code:PA543186
Catalog # PIPA543186
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human GSTT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glutathione S-transferase 1 (theta); Glutathione S-transferase 5; Glutathione S-transferase theta 1; Glutathione S-transferase theta-1; Glutathione transferase T1-1; GST 5-5; GST class-theta-1
Gene Aliases: GSTT1; GSTYRS
UniProt ID: (Human) P30711, (Bovine) Q2NL00, (Rat) Q01579
Entrez Gene ID: (Human) 2952, (Bovine) 517724, (Rat) 25260
Molecular Function:
RNA binding protein
cytoskeletal protein
epimerase/racemase
isomerase
nucleic acid binding
oxidoreductase
reductase
signaling molecule
transferase
translation elongation factor
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.