Invitrogen
Manufacturer Code:MA529314
Catalog # PIMA529314
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 28 |
Immunogen | A synthetic peptide corresponding to the N-terminus of the Human GSTP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: deafness, X-linked 7; epididymis secretory protein Li 22; fatty acid ethyl ester synthase III; Glutathione S-transferase P; GST class-pi; GSTP1 GST3 glutathione S-transferase pi HGNC:4638 DFN7 FAEES3 PI deafness X-linked 7 fatty acid ethyl ester synthase III glutathione transferase glutathione S-transferase pi 1; GSTP1-1
Gene Aliases: DFN7; FAEES3; GST3; GSTP; GSTP1; HEL-S-22; PI
UniProt ID: (Human) P09211
Entrez Gene ID: (Human) 2950
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.