Invitrogen
Manufacturer Code:PA577994
Catalog # PIPA577994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human GSTP1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: deafness, X-linked 7; DFN7 FAEES3 GST3 GSTP PI GSTP1 "deafness X-linked 7 " fatty acid ethyl ester synthase III GST class-pi GSTP1-1 glutathione S-transferase P glutathione S-transferase pi 1; epididymis secretory protein Li 22; fatty acid ethyl ester synthase III; Glutathione S-transferase P; Glutathione S-transferase P 1; GST class-pi; Gst p-1; Gst P1; GST YF-YF; GST-piB; GSTP1-1; Preadipocyte growth factor
Gene Aliases: DFN7; FAEES3; GST3; GSTP; GSTP1; GstpiB; HEL-S-22; PI
UniProt ID: (Human) P09211, (Mouse) P19157
Entrez Gene ID: (Human) 2950, (Mouse) 14870
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.