Novus Biologicals
Manufacturer Code:NBP272371
Catalog # NBP272371
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A12 |
Immunogen | Full length human recombinant protein of human GSTO2(NP_899062) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: bA127L20.1 (novel glutathione-S-transferase); bA127L20.1 bA127L20.1 (novel glutathione-S-transferase) EC 2.5.1.18 glutathione S-transferase omega 2 glutathione S-transferase omega-2 glutathione-S-transferase-like protein GSTO-2; Glutathione S-transferase omega 2-2; Glutathione S-transferase omega-2; Glutathione-dependent dehydroascorbate reductase; glutathione-S-transferase-like protein; GSTO 2-2; GSTO-2; MMA(V) reductase; Monomethylarsonic acid reductase
Gene Aliases: bA127L20.1; GSTO 2-2; GSTO2
UniProt ID: (Human) B4DJW6
Entrez Gene ID: (Human) 119391
Molecular Function:
RNA binding protein
cytoskeletal protein
epimerase/racemase
isomerase
nucleic acid binding
oxidoreductase
reductase
signaling molecule
transferase
translation elongation factor
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.