Invitrogen
Manufacturer Code:PA525736
Catalog # PIPA525736
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 82-110 amino acids from the central region of human GSTO2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA127L20.1 (novel glutathione-S-transferase); Glutathione S-transferase omega 2-2; Glutathione S-transferase omega-2; Glutathione-dependent dehydroascorbate reductase; glutathione-S-transferase-like protein; GSTO 2-2; GSTO 2-2 bA127L20.1 GSTO-2 MMA(V) reductase bA127L20.1 (novel glutathione-S-transferase) glutathione S-transferase omega 2-2 glutathione S-transferase omega-2 glutathione-S-transferase-like protein glutathione-dependent dehydroascorbate reductase monomethylarsonic acid reduc; GSTO-2; MMA(V) reductase; Monomethylarsonic acid reductase
Gene Aliases: 1700020F09Rik; 4930425C18Rik; bA127L20.1; GSTO 2-2; GSTO2
UniProt ID: (Human) B4DJW6, (Mouse) Q8K2Q2
Entrez Gene ID: (Human) 119391, (Mouse) 68214
Molecular Function:
RNA binding protein
cytoskeletal protein
epimerase/racemase
isomerase
nucleic acid binding
oxidoreductase
reductase
signaling molecule
transferase
translation elongation factor
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.