Abnova
Manufacturer Code:MAB9888
Catalog # 89-113-846
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2 |
Immunogen | Recombinant protein corresponding to full length human GSTO1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against full-length recombinant GSTO1.
For Research Use Only
Protein Aliases: DKFZp686H13163; epididymis secretory protein Li 21; Glutathione S-transferase omega 1-1; Glutathione S-transferase omega-1; Glutathione-dependent dehydroascorbate reductase; glutathione-S-transferase like; GSTO 1-1; GSTO-1; GSTTLp28; MMA(V) reductase; Monomethylarsonic acid reductase; P28; S-(Phenacyl)glutathione reductase; SPG-R
Gene Aliases: GSTO 1-1; GSTO1; GSTTLP28; HEL-S-21; P28; SPG-R
UniProt ID: (Human) P78417
Entrez Gene ID: (Human) 9446
Molecular Function:
RNA binding protein
cytoskeletal protein
epimerase/racemase
isomerase
nucleic acid binding
oxidoreductase
reductase
signaling molecule
transferase
translation elongation factor
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.