Invitrogen
Manufacturer Code:MA519027
Catalog # PIMA519027
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B3 |
Immunogen | GSTM5 (NP&lowbar000842 145 a.a. &bsim 218 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: glutathione S-alkyltransferase M5; glutathione S-aralkyltransferase M5; glutathione S-aryltransferase M5; glutathione S-transferase M5; Glutathione S-transferase Mu 5; GST class-mu 5; GSTM5-5; GSTM5-5 GTM5 GST class-mu 5 S-(hydroxyalkyl)glutathione lyase M5 glutathione S-alkyltransferase M5 glutathione S-aralkyltransferase M5 glutathione S-aryltransferase M5 glutathione S-transferase M5 glutathione S-transferase Mu 5; S-(hydroxyalkyl)glutathione lyase M5
Gene Aliases: GSTM5; GSTM5-5; GTM5
UniProt ID: (Human) P46439
Entrez Gene ID: (Human) 2949
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.