Invitrogen
Manufacturer Code:PA580677
Catalog # PIPA580677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human GSTM5/GST class-mu 5 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glutathione S-alkyltransferase M5; glutathione S-aralkyltransferase M5; glutathione S-aryltransferase M5; glutathione S-transferase M5; Glutathione S-transferase Mu 5; GST class-mu 5; GSTM5-5; GSTM5-5 GTM5 GST class-mu 5 S-(hydroxyalkyl)glutathione lyase M5 glutathione S-alkyltransferase M5 glutathione S-aralkyltransferase M5 glutathione S-aryltransferase M5 glutathione S-transferase M5 glutathione S-transferase Mu 5; S-(hydroxyalkyl)glutathione lyase M5
Gene Aliases: GSTM5; GSTM5-5; GTM5
UniProt ID: (Human) P46439
Entrez Gene ID: (Human) 2949
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.