Invitrogen
Manufacturer Code:PA528892
Catalog # PIPA528892
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 218 of Human GSTM5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Fibrous sheath component 2; Fsc2; glutathione S-alkyltransferase M5; glutathione S-aralkyltransferase M5; glutathione S-aryltransferase M5; glutathione S-transferase M5; Glutathione S-transferase Mu 5; glutathione-S-transferase, mu 5; GST class-mu 5; GSTM5-5; GSTM5-5 GTM5 GST class-mu 5 S-(hydroxyalkyl)glutathione lyase M5 glutathione S-alkyltransferase M5 glutathione S-aralkyltransferase M5 glutathione S-aryltransferase M5 glutathione S-transferase M5 glutathione S-transferase Mu 5; S-(hydroxyalkyl)glutathione lyase M5
Gene Aliases: Fsc2; Gstm3; GSTM5; GSTM5-5; GTM5
UniProt ID: (Human) P46439, (Mouse) P48774, (Rat) Q9Z1B2
Entrez Gene ID: (Human) 2949, (Mouse) 14866, (Rat) 64352
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.