Invitrogen
Manufacturer Code:PA5100149
Catalog # PIPA5100149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GSTM4 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110004G14Rik glutathione S-alkyltransferase M4 glutathione S-aralkyltransferase M4 glutathione S-aryltransferase M4 glutathione S-transferase M4 glutathione S-transferase mu 3 glutathione S-transferase Mu 4 glutathione S-transferase mu 4 glutathione-S-transferase mu 4 GST class-mu 4 Gstb4 Gstb-4 GSTM4 GSTM4-4 GSTM7-7 GST-Mu2 GTM4 GTS-Mu2 S-(hydroxyalkyl)glutathione lyase M4 testis tissue sperm-binding protein Li 60n; glutathione S-alkyltransferase M4; glutathione S-aralkyltransferase M4; glutathione S-aryltransferase M4; glutathione S-transferase M4; Glutathione S-transferase Mu 4; Glutathione transferase GSTM7-7; glutathione-S-transferase, mu 4; GST class-mu 4; GST-Mu2; GSTM4-4; GTS-Mu2; Leukotriene C4 synthase GSTM4; S-(hydroxyalkyl)glutathione lyase M4; testis tissue sperm-binding protein Li 60n
Gene Aliases: 1110004G14Rik; Gstb-4; Gstb4; GSTM4; GSTM4-4; Gstm7; GSTM7-7; GTM4
UniProt ID: (Human) Q03013, (Mouse) Q8R5I6
Entrez Gene ID: (Human) 2948, (Mouse) 14865
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.