Novus Biologicals
Manufacturer Code:NBP30453320UL
Catalog # NB095024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-218 of human GSTM4 (NP_000841.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.5.1.18 glutathione S-alkyltransferase M4 glutathione S-aralkyltransferase M4 glutathione S-aryltransferase M4 glutathione S-transferase M4 glutathione S-transferase mu 4 GST class-mu 4 GSTM4-4 GST-Mu2 GTM4 GTS-Mu2 MGC131945 MGC9247 S-(hydroxyalkyl)glutathione lyase M4; glutathione S-alkyltransferase M4; glutathione S-aralkyltransferase M4; glutathione S-aryltransferase M4; glutathione S-transferase M4; Glutathione S-transferase Mu 4; GST class-mu 4; GST-Mu2; GSTM4-4; GTS-Mu2; Leukotriene C4 synthase GSTM4; S-(hydroxyalkyl)glutathione lyase M4; testis tissue sperm-binding protein Li 60n
Gene Aliases: GSTM4; GSTM4-4; GTM4
UniProt ID: (Human) Q03013
Entrez Gene ID: (Human) 2948
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.