Abnova
Manufacturer Code:H00002948A01
Catalog # 89-104-595
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 109 a.a. to 218 a.a) partial recombinant protein with GST tag.,GSTM4 (NP_000841 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant GSTM4.
For Research Use Only
Protein Aliases: glutathione S-alkyltransferase M4; glutathione S-aralkyltransferase M4; glutathione S-aryltransferase M4; glutathione S-transferase M4; Glutathione S-transferase Mu 4; GST class-mu 4; GST-Mu2; GSTM4-4; GTM4; GTS-Mu2; Leukotriene C4 synthase GSTM4; MGC131945; MGC9247; S-(hydroxyalkyl)glutathione lyase M4; testis tissue sperm-binding protein Li 60n
Gene Aliases: GSTM4; GSTM4-4; GTM4
UniProt ID: (Human) Q03013
Entrez Gene ID: (Human) 2948
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.