Proteintech
Manufacturer Code:152141AP150UL
Catalog # 50-173-0070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GSTM3 Fusion Protein Ag7375 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain GST; brain type mu-glutathione S-transferase; Glutathione S transferase Mu 3 GST class mu 3 GST5 GSTB GSTM3 GSTM3 3 GTM3 hGSTM3 3; glutathione S-alkyltransferase M3; glutathione S-aralkyltransferase M3; glutathione S-aryltransferase M3; Glutathione S-transferase GT9.3; glutathione S-transferase M3 (brain); Glutathione S-transferase Mu 3; glutathione S-transferase mu 3 (brain); glutathione S-transferase, Mu-3; GST class-mu 3; GSTM3-3; hGSTM3-3; S-(hydroxyalkyl)glutathione lyase M3
Gene Aliases: Fsc2; GST5; GSTB; GSTM3; GSTM3-3; GTM3; mGSTM5
UniProt ID: (Human) P21266, (Mouse) P19639
Entrez Gene ID: (Human) 2947, (Mouse) 14864
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.