Abnova
Manufacturer Code:H00002939M05
Catalog # 89-022-403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3D4 |
Immunogen | GSTA2 (AAH02895 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant GSTA2.
For Research Use Only
Protein Aliases: glutathione S-alkyltransferase A2; glutathione S-aralkyltransferase A2; glutathione S-aryltransferase A2; Glutathione S-transferase A2; GST class-alpha member 2; GST HA subunit 2; GST-gamma; GST2; GSTA2-2; GTA2; GTH2; liver GST2; MGC10525; S-(hydroxyalkyl)glutathione lyase A2; testis tissue sperm-binding protein Li 59n
Gene Aliases: GST2; GSTA2; GSTA2-2; GTA2; GTH2
UniProt ID: (Human) P09210
Entrez Gene ID: (Human) 2939
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.