Invitrogen
Manufacturer Code:PA521670
Catalog # PIPA521670
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 196 of GSTA2 (Uniprot IDNo.P09210) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glutathione S-alkyltransferase A2; glutathione S-aralkyltransferase A2; glutathione S-aryltransferase A2; Glutathione S-transferase A2; GST class-alpha member 2; GST HA subunit 2; GST-gamma; GST2 GSTA2-2 GTA2 GTH2 GST HA subunit 2 GST class-alpha member 2 GST-gamma S-(hydroxyalkyl)glutathione lyase A2 glutathione S-alkyltransferase A2 glutathione S-aralkyltransferase A2 glutathione S-aryltransferase A2 glutathione S-transferase A2 liver GST2; GSTA2-2; GTH2; liver GST2; S-(hydroxyalkyl)glutathione lyase A2; testis tissue sperm-binding protein Li 59n
Gene Aliases: GST2; GSTA2; GSTA2-2; GTA2; GTH2
UniProt ID: (Human) P09210
Entrez Gene ID: (Human) 2939
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.