Novus Biologicals
Manufacturer Code:NBP30454420UL
Catalog # NB093315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-222 of human GSTA2 (NP_000837.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.5.1.18 glutathione S-aralkyltransferase A2 glutathione S-aryltransferase A2 glutathione S-transferase 2 glutathione S-transferase A2 glutathione S-transferase alpha 2 GST class-alpha member 2 GST HA subunit 2 GST class alpha 2 GST2glutathione S-alkyltransferase A2 GSTA2-2 GST-gamma GTA2 GTH2 HA subunit 2 liver GST2 MGC10525 S-(hydroxyalkyl)glutathione lyase A2; glutathione S-alkyltransferase A2; glutathione S-aralkyltransferase A2; glutathione S-aryltransferase A2; Glutathione S-transferase A2; GST class-alpha member 2; GST HA subunit 2; GST-gamma; GSTA2-2; GTH2; liver GST2; S-(hydroxyalkyl)glutathione lyase A2; testis tissue sperm-binding protein Li 59n
Gene Aliases: GST2; GSTA2; GSTA2-2; GTA2; GTH2
UniProt ID: (Human) P09210
Entrez Gene ID: (Human) 2939
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.