Proteintech
Manufacturer Code:666241IG150UL
Catalog # 50-173-7042
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E1F4 |
Immunogen | GSTA1 Fusion Protein Ag6084 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 13-hydroperoxyoctadecadienoate peroxidase; Androst-5-ene-3,17-dione isomerase; Glutathione S transferase A1 GST class alpha member 1 GST epsilon GST HA subunit 1 GST2 GSTA1 GSTA1 1 GTH1; glutathione S-alkyltransferase A1; glutathione S-aryltransferase A1; glutathione S-transferase 2; Glutathione S-transferase A1; Glutathione S-transferase A1, N-terminally processed; glutathione S-transferase A3 subunit; glutathione S-transferase A5; Glutathione S-transferase alpha M14; Glutathione S-transferase alpha M14, N-terminally processed; Glutathione S-transferase alpha-3; glutathione S-transferase Ha subunit 1; Glutathione S-transferase Ya; Glutathione S-transferase Ya1; Glutathione S-transferase Yc-1; glutathione S-transferase Yc1 subunit; Glutathione-S-transferase alpha type (Ya); GST 2-2; GST A3-3; GST AA; GST class-alpha; GST class-alpha member 1; GST HA subunit 1; GST Yc1; GST-epsilon; GSTA1-1; GTH1; S-(hydroxyalkyl)glutathione lyase A1; testicular tissue protein Li 80
Gene Aliases: GST-epsilon; GST2; Gst2-1; Gsta; GSTA1; GSTA1-1; Gsta3; Gsta5; Gstya; Gstyc1; GTH1; OTTMUSG00000031890; Yc1
UniProt ID: (Human) P08263, (Pig) P51781, (Rat) P04904, (Mouse) P13745
Entrez Gene ID: (Human) 2938, (Pig) 397682, (Rat) 24421, (Mouse) 14857
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.