Invitrogen
Manufacturer Code:PA5102338
Catalog # PIPA5102338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GSS corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI314904 Glutathione synthase glutathione synthetase GS-A/GS-B GSH S GSH synthetase GSHB GSHS GSH-S Gss HEL-S-64p HEL-S-88n MGC14098 OTTHUMP00000030711; Glutathione synthase; Glutathione synthetase; GSH synthetase; GSH-S
Gene Aliases: AI314904; GS-A/GS-B; GSH-S; GSHS; GSS; HEL-S-64p; HEL-S-88n
UniProt ID: (Human) P48637, (Rat) P46413, (Mouse) P51855
Entrez Gene ID: (Human) 2937, (Rat) 25458, (Mouse) 14854
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.