Invitrogen
Manufacturer Code:PA579334
Catalog # PIPA579334
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human Glutathione Reductase recombit protein (Position: K256-R522). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis luminal protein 75; epididymis secretory sperm binding protein Li 122m; glutathione reductase; glutathione reductase 1; Glutathione reductase mitochondrial GR GRase GSR GLUR GRD1; Glutathione reductase, mitochondrial; glutathione S-reductase; GR; GRase
Gene Aliases: AI325518; D8Ertd238e; GLUR; Gr-1; Gr1; GRD1; GSR; HEL-75; HEL-S-122m
UniProt ID: (Human) P00390, (Mouse) P47791
Entrez Gene ID: (Human) 2936, (Mouse) 14782, (Rat) 116686
Molecular Function: dehydrogenase oxidase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.