Novus Biologicals
Manufacturer Code:NBP271768APC
Catalog # NB271768APC
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2G9 |
Immunogen | Full length recombinant protein of human GSPT2 (NP_060564) produced in HEK293T cells. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: eRF3b ERF3BGST2 eukaryotic peptide chain release factor GTP-binding subunit ERF3B Eukaryotic peptide chain release factor subunit 3b FLJ10441 G1 to S phase transition 2 G1 to S phase transition protein 2 homolog; Eukaryotic peptide chain release factor GTP-binding subunit ERF3B; Eukaryotic peptide chain release factor subunit 3b; G1 to S phase transition protein 2 homolog
Gene Aliases: ERF3B; GSPT2; GST2
UniProt ID: (Human) Q8IYD1
Entrez Gene ID: (Human) 23708
Molecular Function:
G-protein
RNA binding protein
enzyme modulator
hydrolase
nucleic acid binding
translation elongation factor
translation factor
translation initiation factor
translation release factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.