Novus Biologicals
Manufacturer Code:NBP247442
Catalog # NBP247442
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: GGAANNHGAGSGAGGRAAPVESSQEEQSLCEGSNSAVS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ERF3A eRF3aFLJ38048 ETF3A eukaryotic peptide chain release factor GTP-binding subunit ERF3A Eukaryotic peptide chain release factor subunit 3a FLJ39067 G1 to S phase transition 1551G9.2 G1 to S phase transition protein 1 homolog GST1; Eukaryotic peptide chain release factor GTP-binding subunit ERF3A; Eukaryotic peptide chain release factor subunit 3a; G1 to S phase transition protein 1 homolog
Gene Aliases: 551G9.2; ERF3A; ETF3A; GSPT1; GST1
UniProt ID: (Human) P15170
Entrez Gene ID: (Human) 2935
Molecular Function:
G-protein
RNA binding protein
enzyme modulator
hydrolase
nucleic acid binding
translation elongation factor
translation factor
translation initiation factor
translation release factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.